COMH042374C5 Combi-Blocks, Inc.

1-Pyrrolidinecarboxylic acid, 3-(hydroxymethyl)-4-(3-methoxyphenyl)-, 1,1-dimethylethyl ester, (3s,4r)-

  • MDL number MFCD27981367
  • Cas Number : 2173052-95-2
  • Molecular Weight : 307.39

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-1048

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-1048
Availability details for
Quantity
  • Availability
  • Add to Cart