COMH042374EF Combi-Blocks, Inc.

2-(4-Ethylphenyl)-3-methylquinoline-4-carboxylic acid

  • MDL number MFCD01993649
  • Cas Number : 350997-48-7
  • Molecular Weight : 291.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-1090

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-1090
Availability details for
Quantity
  • Availability
  • Add to Cart