COMH0423750C Combi-Blocks, Inc.

3-(1,3-Dioxo-1,3-dihydro-2h-pyrrolo[3,4-c]pyridin-2-yl)propanoic acid

  • MDL number MFCD03767403
  • Cas Number : 681837-30-9
  • Molecular Weight : 220.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-1119

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-1119
Availability details for
Quantity
  • Availability
  • Add to Cart