COMH04237526 Combi-Blocks, Inc.

2-(1-Naphthyl)quinoline-4-carboxylic acid

  • MDL number MFCD01106639
  • Cas Number : 6265-23-2
  • Molecular Weight : 299.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-1145

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-1145
Availability details for
Quantity
  • Availability
  • Add to Cart