COMH04237539 Combi-Blocks, Inc.

2-Mercapto-3,5,6-trimethylthieno[2,3-d]pyrimidin-4(3h)-one

  • MDL number MFCD01907117
  • Cas Number : 59898-59-8
  • Molecular Weight : 226.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-1164

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-1164
Availability details for
Quantity
  • Availability
  • Add to Cart