COMH042375BB Combi-Blocks, Inc.

4-[(2-Chloropropanoyl)amino]benzamide

  • MDL number MFCD06336117
  • Cas Number : 749907-03-7
  • Molecular Weight : 226.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-1295

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-1295
Availability details for
Quantity
  • Availability
  • Add to Cart