COMH042375D7 Combi-Blocks, Inc.

2-Cyano-n-(4-ethoxyphenyl)acetamide

  • MDL number MFCD02194318
  • Cas Number : 51838-07-4
  • Molecular Weight : 204.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-1324

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-1324
Availability details for
Quantity
  • Availability
  • Add to Cart