COMH04237608 Combi-Blocks, Inc.

1-(2-Sec-butyl-phenyl)-5-oxo-pyrrolidine-3-carboxylic acid

  • MDL number MFCD04606975
  • Cas Number : 556007-05-7
  • Molecular Weight : 261.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-1374

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-1374
Availability details for
Quantity
  • Availability
  • Add to Cart