COMH04237615 Combi-Blocks, Inc.

Methyl 4-nitrooxanthrene-2-carboxylate

  • MDL number MFCD08282888
  • Cas Number : 924843-92-5
  • Molecular Weight : 287.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-1387

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-1387
Availability details for
Quantity
  • Availability
  • Add to Cart