COMH04237648 Combi-Blocks, Inc.

2-Amino-6-phenyl-4,5,6,7-tetrahydro-1-benzothiophene-3-carbonitrile

  • MDL number MFCD02656574
  • Cas Number : 37071-21-9
  • Molecular Weight : 254.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-1438

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-1438
Availability details for
Quantity
  • Availability
  • Add to Cart