COMH0423764D Combi-Blocks, Inc.

2-[(Chloroacetyl)amino]-4-ethyl-5-methylthiophene-3-carboxamide

  • MDL number MFCD03380901
  • Cas Number : 544657-74-1
  • Molecular Weight : 260.74

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-1443

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-1443
Availability details for
Quantity
  • Availability
  • Add to Cart