COMH04237650 Combi-Blocks, Inc.

N-[(4-Chlorophenyl)sulfonyl]-n-(4-methylphenyl)glycine

  • MDL number MFCD02813995
  • Cas Number : 896897-91-9
  • Molecular Weight : 339.8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-1446

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-1446
Availability details for
Quantity
  • Availability
  • Add to Cart