COMH04237657 Combi-Blocks, Inc.

3-(3,5-Difluorophenyl)-7-hydroxy-2h-chromen-2-one

  • MDL number MFCD09854772
  • Cas Number : 951957-81-6
  • Molecular Weight : 274.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-1453

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-1453
Availability details for
Quantity
  • Availability
  • Add to Cart