COMH04237674 Combi-Blocks, Inc.

2-(4-tert-Butylphenoxy)butanoyl chloride

  • MDL number MFCD12197839
  • Cas Number : 679837-26-4
  • Molecular Weight : 254.76

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-1482

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-1482
Availability details for
Quantity
  • Availability
  • Add to Cart