COMH04237688 Combi-Blocks, Inc.

N-(4-Nitrobenzoyl)glutamic acid

  • MDL number MFCD00066818
  • Cas Number : 22536-03-4
  • Molecular Weight : 296.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-1502

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-1502
Availability details for
Quantity
  • Availability
  • Add to Cart