COMH04237690 Combi-Blocks, Inc.

5-Bromo-2,2-dimethyl-6-nitro-1,3-benzodioxole

  • MDL number MFCD01463448
  • Cas Number : 342043-80-5
  • Molecular Weight : 274.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-1510

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-1510
Availability details for
Quantity
  • Availability
  • Add to Cart