COMH042376A3 Combi-Blocks, Inc.

2-(2,4-Dichlorophenyl)quinoline-4-carbohydrazide

  • MDL number MFCD02375270
  • Cas Number : 524932-68-1
  • Molecular Weight : 332.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-1529

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-1529
Availability details for
Quantity
  • Availability
  • Add to Cart