COMH042376A5 Combi-Blocks, Inc.

Dimethyl 2,6-dimethyl-4-(3-nitrophenyl)pyridine-3,5-dicarboxylate

  • MDL number MFCD04222132
  • Cas Number : 76258-20-3
  • Molecular Weight : 344.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-1531

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-1531
Availability details for
Quantity
  • Availability
  • Add to Cart