COMH042376B8 Combi-Blocks, Inc.

4-Chloro-3-([(2,3-dimethylphenyl)amino]sulfonyl)benzoic acid

  • MDL number MFCD05812470
  • Cas Number : 884990-43-6
  • Molecular Weight : 339.8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-1550

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-1550
Availability details for
Quantity
  • Availability
  • Add to Cart