COMH042376C4 Combi-Blocks, Inc.

2-Chloro-5-([(chloroacetyl)amino]methyl)benzoic acid

  • MDL number MFCD02625662
  • Cas Number : 91587-41-6
  • Molecular Weight : 262.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-1562

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-1562
Availability details for
Quantity
  • Availability
  • Add to Cart