COMH042376D6 Combi-Blocks, Inc.

2-[4-(Diethylamino)-2-methylphenyl]-6-methyl-2h-1,2,3-benzotriazol-5-amine

  • MDL number MFCD05796782
  • Cas Number : 708287-34-7
  • Molecular Weight : 309.41

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-1580

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-1580
Availability details for
Quantity
  • Availability
  • Add to Cart