COMH0423771E Combi-Blocks, Inc.

4-Butyl-5-hydroxy-7-methyl-2h-chromen-2-one

  • MDL number MFCD03712848
  • Cas Number : 690680-03-6
  • Molecular Weight : 232.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-1652

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-1652
Availability details for
Quantity
  • Availability
  • Add to Cart