COMH04237720 Combi-Blocks, Inc.

2-(4-Chlorophenyl)-8-methylquinoline-4-carbohydrazide

  • MDL number MFCD03420457
  • Cas Number : 590359-89-0
  • Molecular Weight : 311.77

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-1654

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-1654
Availability details for
Quantity
  • Availability
  • Add to Cart