COMH04237753 Combi-Blocks, Inc.

Methyl 6-nitropyrido[1,2-a]benzimidazole-8-carboxylate

  • MDL number MFCD04061736
  • Cas Number : 696628-83-8
  • Molecular Weight : 271.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-1705

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-1705
Availability details for
Quantity
  • Availability
  • Add to Cart