COMH04237781 Combi-Blocks, Inc.

2-(2-Methylphenyl)quinoline-4-carbohydrazide

  • MDL number MFCD02612351
  • Cas Number : 438197-10-5
  • Molecular Weight : 277.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-1752

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-1752
Availability details for
Quantity
  • Availability
  • Add to Cart