COMH042377AE Combi-Blocks, Inc.

N-(3,4-Dimethylphenyl)-n'-(6-methyl-4-oxo-1,4-dihydropyrimidin-2-yl)guanidine

  • MDL number MFCD19103549
  • Cas Number : 874006-44-7
  • Molecular Weight : 271.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-1798

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-1798
Availability details for
Quantity
  • Availability
  • Add to Cart