COMH042377B9 Combi-Blocks, Inc.

1-(2,6-Dichlorobenzyl)-3,5-dimethyl-1h-pyrazol-4-amine

  • MDL number MFCD02055613
  • Cas Number : 898055-78-2
  • Molecular Weight : 270.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-1809

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-1809
Availability details for
Quantity
  • Availability
  • Add to Cart