COMH042377D5 Combi-Blocks, Inc.

1-Isothiocyanato-2,3-dimethoxybenzene

  • MDL number MFCD05668170
  • Cas Number : 80143-14-2
  • Molecular Weight : 195.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-1837

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-1837
Availability details for
Quantity
  • Availability
  • Add to Cart