COMH04237802 Combi-Blocks, Inc.

Ethyl 2-amino-5-[(isopropylamino)carbonyl]-4-methylthiophene-3-carboxylate

  • MDL number MFCD07617180
  • Cas Number : 886672-89-5
  • Molecular Weight : 270.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-1884

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-1884
Availability details for
Quantity
  • Availability
  • Add to Cart