COMH0423780F Combi-Blocks, Inc.

N-(4-Acetylphenyl)-1-benzothiophene-3-carboxamide

  • MDL number MFCD03419978
  • Cas Number : 886629-26-1
  • Molecular Weight : 295.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-1898

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-1898
Availability details for
Quantity
  • Availability
  • Add to Cart