COMH0423782D Combi-Blocks, Inc.

N-Methyl-n-(4-pyrrolidin-1-ylbenzyl)amine

  • MDL number MFCD07686040
  • Cas Number : 823188-79-0
  • Molecular Weight : 190.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-1928

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-1928
Availability details for
Quantity
  • Availability
  • Add to Cart