COMH0423783A Combi-Blocks, Inc.

3-(2,3,4-Trimethoxyphenyl)pentanedioic acid

  • MDL number MFCD10037577
  • Cas Number : 948007-37-2
  • Molecular Weight : 298.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-1941

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-1941
Availability details for
Quantity
  • Availability
  • Add to Cart