COMH04237861 Combi-Blocks, Inc.

5-Bromo-2-[(4-methylbenzyl)oxy]benzaldehyde

  • MDL number MFCD02611658
  • Cas Number : 426224-20-6
  • Molecular Weight : 305.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-1980

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-1980
Availability details for
Quantity
  • Availability
  • Add to Cart