COMH0423786C Combi-Blocks, Inc.

N-(3-Methoxypropyl)-2-nitrobenzamide

  • MDL number MFCD01212155
  • Cas Number : 347907-34-0
  • Molecular Weight : 238.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-1991

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-1991
Availability details for
Quantity
  • Availability
  • Add to Cart