COMH0423796A Combi-Blocks, Inc.

5-(Azepan-1-ylsulfonyl)-2-methylbenzoic acid

  • MDL number MFCD04443695
  • Cas Number : 637320-93-5
  • Molecular Weight : 297.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-2248

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-2248
Availability details for
Quantity
  • Availability
  • Add to Cart