COMH04237979 Combi-Blocks, Inc.

5-[(4-Chloro-3-methylphenoxy)methyl]-4-methyl-4h-1,2,4-triazole-3-thiol

  • MDL number MFCD02934387
  • Cas Number : 447412-27-3
  • Molecular Weight : 269.75

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-2263

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-2263
Availability details for
Quantity
  • Availability
  • Add to Cart