COMH04237980 Combi-Blocks, Inc.

N-(4-Ethylphenyl)-n-(methylsulfonyl)alanine

  • MDL number MFCD13032121
  • Cas Number : 1049806-38-3
  • Molecular Weight : 271.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-2270

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-2270
Availability details for
Quantity
  • Availability
  • Add to Cart