COMH04237985 Combi-Blocks, Inc.

Ethyl 2-[(cyanoacetyl)amino]-4-phenylthiophene-3-carboxylate

  • MDL number MFCD03394771
  • Cas Number : 546090-58-8
  • Molecular Weight : 314.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-2275

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-2275
Availability details for
Quantity
  • Availability
  • Add to Cart