COMH042379B4 Combi-Blocks, Inc.

5-Amino-2-(4-methoxyphenyl)-2h-benzotriazole

  • MDL number MFCD00434232
  • Cas Number : 6659-92-3
  • Molecular Weight : 240.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-2323

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-2323
Availability details for
Quantity
  • Availability
  • Add to Cart