COMH042379BC Combi-Blocks, Inc.

N-(2,4-Dimethylphenyl)-n'-(6-methyl-4-oxo-1,4-dihydropyrimidin-2-yl)guanidine

  • MDL number MFCD19103547
  • Cas Number : 370871-14-0
  • Molecular Weight : 271.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-2331

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-2331
Availability details for
Quantity
  • Availability
  • Add to Cart