COMH042379D2 Combi-Blocks, Inc.

1-[(4-Isothiocyanatophenyl)sulfonyl]-3,5-dimethylpiperidine

  • MDL number MFCD09971951
  • Cas Number : 956576-69-5
  • Molecular Weight : 310.44

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-2353

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-2353
Availability details for
Quantity
  • Availability
  • Add to Cart