COMH042379F5 Combi-Blocks, Inc.

3,4-Dihydrospiro[1-benzopyran-2,1'-cyclopentane]-4-amine

  • MDL number MFCD10006200
  • Cas Number : 70505-77-0
  • Molecular Weight : 203.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-2388

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-2388
Availability details for
Quantity
  • Availability
  • Add to Cart