COMH04237A17 Combi-Blocks, Inc.

1-Benzyl-3,4-dimethyl-pyrrolidine-3,4-dicarboxylic acid

  • MDL number MFCD05662760
  • Cas Number : 885956-54-7
  • Molecular Weight : 277.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-2422

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-2422
Availability details for
Quantity
  • Availability
  • Add to Cart