COMH04237A24 Combi-Blocks, Inc.

4-Ethyl-5-[1-(ethylsulfonyl)piperidin-3-yl]-4h-1,2,4-triazole-3-thiol

  • MDL number MFCD08569904
  • Cas Number : 932918-75-7
  • Molecular Weight : 304.44

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-2435

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-2435
Availability details for
Quantity
  • Availability
  • Add to Cart