COMH04237A2D Combi-Blocks, Inc.

5-(4-Methoxybenzyl)-4-methyl-4h-1,2,4-triazole-3-thiol

  • MDL number MFCD02091587
  • Cas Number : 69198-26-1
  • Molecular Weight : 235.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-2444

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-2444
Availability details for
Quantity
  • Availability
  • Add to Cart