COMH04237A75 Combi-Blocks, Inc.

N,N-Dimethylazetidine-3-carboxamide 2,2,2-trifluoroacetate

  • MDL number MFCD29034959
  • Cas Number : 927390-60-1
  • Molecular Weight : 242.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-2517

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-2517
Availability details for
Quantity
  • Availability
  • Add to Cart