COMH04237A91 Combi-Blocks, Inc.

2-(3-Chlorophenyl)quinoline-4-carbohydrazide

  • MDL number MFCD03420728
  • Cas Number : 886628-78-0
  • Molecular Weight : 297.74

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-2545

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-2545
Availability details for
Quantity
  • Availability
  • Add to Cart