COMH04237AA7 Combi-Blocks, Inc.

N-(4-Chlorophenyl)-2-hydrazino-2-oxoacetamide

  • MDL number MFCD00856252
  • Cas Number : 53117-27-4
  • Molecular Weight : 213.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-2567

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-2567
Availability details for
Quantity
  • Availability
  • Add to Cart