COMH04237AA9 Combi-Blocks, Inc.

5-(Chloromethyl)-2-(3-methoxyphenyl)-1,3-thiazole hydrochloride

  • MDL number MFCD18071249
  • Cas Number : 915920-12-6
  • Molecular Weight : 276.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-2569

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-2569
Availability details for
Quantity
  • Availability
  • Add to Cart