COMH04237AB2 Combi-Blocks, Inc.

6-(Chloromethyl)-2-cyclopropylpyrimidin-4-ol

  • MDL number MFCD00084963
  • Cas Number : 94171-07-0
  • Molecular Weight : 184.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-2578

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QZ-2578
Availability details for
Quantity
  • Availability
  • Add to Cart